2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine

A193262-100MG A193262-250MG A193262-1G A193262-5G
77236-960EA 25.75 USD
77236-960 77236-962 77550-244 77567-802
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine 97%
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine

Precaución: For research use only.

Formula: C₁₂H₆F₅N
MW: 259.18 g/mol
MDL Number: MFCD28144542
CAS Number: 387827-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR