3,5-Difluoro-4-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A193241-100G A193241-10G A193241-250MG A193241-25G A193241-5G
77274-530EA 267 USD
77274-530 77274-532 77274-534 77748-229 77748-230
3,5-Difluoro-4-methoxybenzoic acid 98%
3,5-Difluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MDL Number: MFCD01090994
CAS Number: 319-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR