4,4',4''-(1,3,5-Triazine-2,4,6-triyl)tribenzaldehyde 97%

Supplier: Ambeed
Warning

A193146-100MG A193146-1G A193146-250MG
77260-540EA 14 USD
77260-540 77260-542 77260-544
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)tribenzaldehyde 97%
4,4',4''-(1,3,5-Triazine-2,4,6-triyl)tribenzaldehyde

Precaución: for Research Use Only

Formula: C₂₄H₁₅N₃O₃
MDL Number: MFCD31631359
CAS Number: 443922-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR