5-Fluoro-2-iodoaniline 97%

Supplier: Ambeed
Warning

A193139-100G A193139-10G A193139-1G A193139-250MG A193139-25G A193139-5G
77287-508EA 479.98 USD
77287-508 77287-510 77287-512 77287-514 77287-516 77287-518
5-Fluoro-2-iodoaniline 97%
5-Fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD02093953
CAS Number: 255724-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR