3-Methyl-4-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-nitrobenzoic acid, 4-Nitro-m-toluic acid

A193041-1000G A193041-100G A193041-25G A193041-500G
77236-948EA 63 USD
77236-948 77236-950 77236-952 77236-954
3-Methyl-4-nitrobenzoic acid 97%
3-Methyl-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00007168
CAS Number: 3113-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR