Isobutyramide 97%

Supplier: Ambeed
Warning

A192694-100G A192694-25G A192694-5G
77274-524EA 11 USD
77274-524 77274-526 77274-528
Isobutyramide 97%
Isobutyramide

Precaución: for Research Use Only

Formula: C₄H₉NO
MDL Number: MFCD00008019
CAS Number: 563-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR