3-[4-(dimethylamino)phenyl]propanoic acid 97%

Supplier: Ambeed
Warning

A192689-1G A192689-250MG A192689-25G A192689-5G
77274-516EA 35.02 USD
77274-516 77274-518 77274-520 77274-522
3-[4-(dimethylamino)phenyl]propanoic acid 97%
3-[4-(dimethylamino)phenyl]propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.24 g/mol
MDL Number: MFCD06823974
CAS Number: 73718-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR