2,4-Dimethylbenzonitrile 98%

Supplier: Ambeed
Warning

A192454-1G A192454-250MG
77248-592EA 19.57 USD
77248-592 77248-594
2,4-Dimethylbenzonitrile 98%
2,4-Dimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
MDL Number: MFCD00016377
CAS Number: 21789-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR