3-Aminopropionitrile ≥98% stabilized

Supplier: Ambeed
Danger

Synonyms: β-Aminopropionitrile, 3-Aminopropanenitrile

Stabilization: Stabilised with potassium carbonate 0.1%

A192419-1G A192419-25G A192419-5G A192419-10G
77274-492EA 5.15 USD
77274-492 77274-494 77274-496 77567-792
3-Aminopropionitrile ≥98% stabilized
3-Aminopropionitrile

Precaución: For research use only.

Formula: C₃H₆N₂
MW: 70.10 g/mol
MDL Number: MFCD00014820
CAS Number: 151-18-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR