Methyl-3-methyl-2-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-methyl-2-nitrobenzoate

A192298-1000G A192298-100G A192298-10G A192298-25G A192298-500G
77274-472EA 147 USD
77274-472 77274-474 77274-476 77274-478 77274-480 77274-482
Methyl-3-methyl-2-nitrobenzoate 97%
Methyl 3-methyl-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD01003947
CAS Number: 5471-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR