3'-(Trifluoromethyl)propiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-one, m-(Trifluoromethyl)propiophenone

A192273-1G A192273-25G A192273-5G A192273-100G
77274-466EA 5.15 USD
77274-466 77274-468 77274-470 77748-227
3'-(Trifluoromethyl)propiophenone 98%
3'-(Trifluoromethyl)propiophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
MDL Number: MFCD00039227
CAS Number: 1533-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR