PRX-08066 98+%

Supplier: Ambeed
Warning

Synonyms: 5-((4-((6-Chlorothieno[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)methyl)-2-fluorobenzonitrile

A192115-1MG A192115-5MG
77236-934EA 29.87 USD
77236-934 77236-936
PRX-08066 98+%
PRX-08066

Precaución: for Research Use Only

Formula: C₁₉H₁₇ClFN₅S
MW: 401.89 g/mol
MDL Number: MFCD20488005
CAS Number: 866206-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR