2-Amino-5-bromo-4-methylthiazole hydrochloride 98%

Supplier: Ambeed
Warning

A191883-1G A191883-25G A191883-5G
77248-584EA 25.75 USD
77248-584 77248-586 77248-588
2-Amino-5-bromo-4-methylthiazole hydrochloride 98%
2-Amino-5-bromo-4-methylthiazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₆BrClN₂S
MW: 229.53 g/mol
MDL Number: MFCD09265523
CAS Number: 133692-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR