4,5-Difluoro-2-methoxybenzonitrile 98%
Supplier: Ambeed
|
Danger
|
A191836-1G
A191836-250MG
77287-500EA
35.02
USD
77287-500
77287-502
4,5-Difluoro-2-methoxybenzonitrile 98%
4,5-Difluoro-2-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅F₂NO MW: 169.13 g/mol |
MDL Number:
MFCD06660346 CAS Number: 425702-28-9 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...