1,4-Di(1H-pyrazol-4-yl)benzene 98%

Supplier: Ambeed
Warning

A191342-5G A191342-1G
77236-920EA 456.29 USD
77236-920 77550-222
1,4-Di(1H-pyrazol-4-yl)benzene 98%
1,4-Di(1H-pyrazol-4-yl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄
MW: 210.23 g/mol
MDL Number: MFCD23704797
CAS Number: 1036248-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR