5-((5-(2-(3-Aminopropoxy)-6-methoxyphenyl)-1H-pyrazol-3-yl)amino)pyrazine-2-carbonitrile dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
A191306-10MG
A191306-1MG
A191306-25MG
A191306-2MG
A191306-50MG
A191306-5MG
77236-908EA
99
USD
77236-908
77236-910
77236-912
77236-914
77236-916
77236-918
5-((5-(2-(3-Aminopropoxy)-6-methoxyphenyl)-1H-pyrazol-3-yl)amino)pyrazine-2-carbonitrile dihydrochloride 98%
5-((5-(2-(3-Aminopropoxy)-6-methoxyphenyl)-1H-pyrazol-3-yl)amino)pyrazine-2-carbonitrile dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₈H₂₁Cl₂N₇O₂ |
MDL Number:
MFCD27918637 CAS Number: 1234015-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...