3-Dimethylamino-2-methylpropyl chloride hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-N,N,2-trimethylpropan-1-amine hydrochloride

A191096-5G A191096-25G
77274-430EA 15 USD
77274-430 77748-223
3-Dimethylamino-2-methylpropyl chloride hydrochloride 98%
3-Dimethylamino-2-methylpropyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅Cl₂N
MDL Number: MFCD00012514
CAS Number: 4261-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR