3-Aminophenethyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Aminophenyl)ethanol

A190825-10G A190825-1G A190825-250MG A190825-5G
77274-420EA 315 USD
77274-420 77274-422 77274-424 77274-426
3-Aminophenethyl alcohol 97%
3-Aminophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD02683213
CAS Number: 52273-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR