2-(Benzylthio)-9H-purin-6-amine 97%

Supplier: Ambeed
Warning

A190800-100MG A190800-1G A190800-250MG
77260-504EA 9.27 USD
77260-504 77260-506 77260-508
2-(Benzylthio)-9H-purin-6-amine 97%
2-(Benzylthio)-9H-purin-6-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N₅S
MW: 257.31 g/mol
MDL Number: MFCD30729982
CAS Number: 64542-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR