1H-Benzotriazole-1-methanol 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Hydroxymethyl)benzotriazole

A190585-100G A190585-1G A190585-25G A190585-500G A190585-5G A190585-10G
77236-874EA 113.3 USD
77236-874 77236-876 77236-878 77236-880 77236-882 77615-614
1H-Benzotriazole-1-methanol 95%
1H-Benzotriazole-1-methanol

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MW: 149.1500 g/mol
MDL Number: MFCD00179118
CAS Number: 28539-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR