1-(4-((2-Aminopyrimidin-4-yl)oxy)phenyl)-3-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)urea 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
AUZ 454
A190561-10MG
A190561-1MG
A190561-5MG
A190561-50MG
A190561-100MG
77260-498EA
40.17
USD
77260-498
77260-500
77260-502
77615-612
77748-218
1-(4-((2-Aminopyrimidin-4-yl)oxy)phenyl)-3-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)urea 98+%
1-(4-((2-Aminopyrimidin-4-yl)oxy)phenyl)-3-(4-((4-methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)urea
Precaución: for Research Use Only
Formula:
C₂₄H₂₆F₃N₇O₂ MW: 501.5 g/mol |
MDL Number:
MFCD19443769 CAS Number: 853299-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...