4-Chloro-5-methoxypyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

A190462-1G A190462-25G A190462-5G
77287-470EA 20 USD
77287-470 77287-472 77287-474
4-Chloro-5-methoxypyridazin-3(2H)-one 98%
4-Chloro-5-methoxypyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MDL Number: MFCD04971392
CAS Number: 63910-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR