2-Aminothiazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Thiazol-2-amine hydrochloride, 2-Thiazolamine hydrochloride

A190241-10G A190241-25G A190241-5G
77260-486EA 10.3 USD
77260-486 77260-488 77260-490
2-Aminothiazole hydrochloride 95%
2-Aminothiazole hydrochloride

Precaución: for Research Use Only

Formula: C₃H₄N₂S^·HCl
MW: 136.6 g/mol
MDL Number: MFCD00270826
CAS Number: 6142-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR