Methyl (E)-3-(3-aminophenyl)acrylate 97%

Supplier: Ambeed
Warning

A190150-250MG A190150-1G
77236-856EA 54.59 USD
77236-856 77746-218
Methyl (E)-3-(3-aminophenyl)acrylate 97%
Methyl (E)-3-(3-aminophenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
MDL Number: MFCD20527968
CAS Number: 163487-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR