3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Carbamoylpiperazine

A190032-100MG A190032-1G A190032-250MG A190032-5G
77287-462EA 25.75 USD
77287-462 77287-464 77287-466 77748-217
3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester 98%
3-Carbamoylpiperazine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₉N₃O₃
MW: 229.28 g/mol
MDL Number: MFCD09608050
CAS Number: 112257-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR