Potassium trifluoro(propyl)borate 97%

Supplier: Ambeed
Danger

A189910-1G A189910-5G
77248-540EA 25 USD
77248-540 77593-212
Potassium trifluoro(propyl)borate 97%
Potassium trifluoro(propyl)borate

Precaución: for Research Use Only

Formula: C₃H₇BF₃K
MDL Number: MFCD09992883
CAS Number: 897067-94-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR