4-Methyl-3-phenylpiperazin-2-one 97%

Supplier: Ambeed
Warning

A189757-100MG A189757-250MG
77236-840EA 27.81 USD
77236-840 77236-842
4-Methyl-3-phenylpiperazin-2-one 97%
4-Methyl-3-phenylpiperazin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O
MW: 190.24 g/mol
MDL Number: MFCD17019390
CAS Number: 5368-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR