Pexmetinib ≥98%

Supplier: Ambeed
Warning

Synonyms: ARRY614, (Z)-N-(3-(tert-butyl)-1-(p-tolyl)-1H-pyrazol-5-yl)-N'-(5-fluoro-2-((1-(2-hydroxyethyl)-1H-indazol-5-yl)oxy)benzyl)carbamimidic acid, ARRY-614, Pexmetinib, 1-(3-(tert-butyl)-1-(p-tolyl)-1H-pyrazol-5-yl)-3-(5-fluoro-2-((1-(2-hydroxyethyl)-1H-indazol-5-yl)oxy)benzyl)urea, ARRY 614, 1-[5-tert-butyl-2-(4-methylphenyl)pyrazol-3-yl]-3-[[5-fluoro-2-[1-(2-hydroxyethyl)indazol-5-yl]oxyphenyl]methyl]urea

A189540-10MG A189540-1MG A189540-25MG A189540-5MG
77260-466EA 116.39 USD
77260-466 77260-468 77260-470 77260-472
Pexmetinib ≥98%
Pexmetinib

Precaución: for Research Use Only

Formula: C₃₁H₃₃FN₆O₃
MW: 556.63 g/mol
MDL Number: MFCD28502055
CAS Number: 945614-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR