2,2,2-Trifluoroethyl nonafluorobutanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulphonate, 2,2,2-Trifluoroethyl Perfluorobutanesulphonate, Nonafluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester, Perfluorobutanesulphonic acid 2,2,2-Trifluoroethyl Ester

A189378-1G A189378-5G
77248-524EA 20 USD
77248-524 77248-526
2,2,2-Trifluoroethyl nonafluorobutanesulfonate 98%
2,2,2-Trifluoroethyl nonafluorobutanesulfonate

Precaución: for Research Use Only

Formula: C₆H₂F₁₂O₃S
MDL Number: MFCD03093638
CAS Number: 79963-95-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR