S-Phenyl-L-cysteine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(phenylthio)propanoic acid, H-Cys(phenyl)-OH

A189316-100G A189316-100MG A189316-1G A189316-250MG A189316-25G A189316-5G
77248-510EA 130 USD
77248-510 77248-512 77248-514 77248-516 77248-518 77248-520
S-Phenyl-L-cysteine 97%
S-Phenyl-L-cysteine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂S
MDL Number: MFCD01318758
CAS Number: 34317-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR