4-Methyl-3-(trifluoromethyl)benzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)-1-methyl-2-(trifluoromethyl)benzene

A189313-100MG A189313-1G A189313-250MG A189313-5G
77287-452EA 39.14 USD
77287-452 77287-454 77287-456 77287-458
4-Methyl-3-(trifluoromethyl)benzyl bromide 95%
4-Methyl-3-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₉H₈BrF₃
MW: 253.06 g/mol
MDL Number: MFCD01631612
CAS Number: 261952-19-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR