BH3I-1 98%

Supplier: Ambeed
Warning

Synonyms: 5-[(4-bromophenyl)methylene]-a-(1-methylethyl)-4-oxo-2-thioxo-3-thiazolidineacetic acid, 2-(5-(4-Bromobenzylidene)-4-oxo-2-thioxothiazolidin-3-yl)-3-methylbutanoic acid

A189286-10MG A189286-1MG A189286-25MG A189286-50MG A189286-5MG
77236-824EA 206 USD
77236-824 77236-826 77236-828 77236-830 77236-832
BH3I-1 98%
BH3I-1

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrNO₃S₂
MW: 400.31 g/mol
MDL Number: MFCD03453544
CAS Number: 300817-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR