4-Fluoro-2-(trifluoromethyl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyano-5-fluorobenzotrifluoride
A189201-10G
A189201-25G
A189201-5G
A189201-100G
77274-380EA
8
USD
77274-380
77274-382
77274-384
77567-764
4-Fluoro-2-(trifluoromethyl)benzonitrile 98%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Precaución: For research use only.
Formula:
C₈H₃F₄N MW: 189.11 g/mol |
MDL Number:
MFCD00061283 CAS Number: 194853-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...