trans-4-Chlorocinnamic-acid 95%

Supplier: Ambeed
Warning

Synonyms: (2E)-3-(4-Chlorophenyl)prop-2-enoic acid, (2E)-3-(4-Chlorophenyl)acrylic acid, trans-3-(4-Chlorophenyl)acrylic acid, (E)-3-(4-Chlorophenyl)acrylic acid

A189179-100G A189179-10G A189179-25G A189179-500G A189179-5G
77236-814EA 38.11 USD
77236-814 77236-816 77236-818 77236-820 77236-822
trans-4-Chlorocinnamic-acid 95%
trans-4-Chlorocinnamic-acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
MW: 182.60 g/mol
MDL Number: MFCD00004396
CAS Number: 940-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR