3-(Benzyloxy)-2,2-dimethylpropan-1-ol 95%

Supplier: Ambeed
Warning

A189134-1G A189134-250MG A189134-5G
77236-808EA 120.51 USD
77236-808 77236-810 77236-812
3-(Benzyloxy)-2,2-dimethylpropan-1-ol 95%
3-(Benzyloxy)-2,2-dimethylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
MDL Number: MFCD00233351
CAS Number: 66582-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR