N-(4-Methyl-3-((4-(pyridin-3-yl)thiazol-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulfonate 98+%

Supplier: Ambeed
Warning

Synonyms: Masitinib mesylate

A189077-10MG A189077-1MG A189077-50MG A189077-5MG A189077-250MG
77236-800EA 28.84 USD
77236-800 77236-802 77236-804 77236-806 77746-212
N-(4-Methyl-3-((4-(pyridin-3-yl)thiazol-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulfonate 98+%
N-(4-Methyl-3-((4-(pyridin-3-yl)thiazol-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulfonate

Precaución: for Research Use Only

Formula: C₂₉H₃₄N₆O₄S₂
MW: 594.75 g/mol
MDL Number: MFCD12546334
CAS Number: 1048007-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR