Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4,5,6,7-tetrahydro-1H-indazole-3-carboxylate
A188676-1G
A188676-250MG
77287-448EA
54.59
USD
77287-448
77287-450
Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate 98%
Ethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂O₂ MW: 194.24 g/mol |
MDL Number:
MFCD02942367 CAS Number: 4492-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...