4'-Hydroxy-2'-methylacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 4′-Hydroxy-2′-methylacetophenone, 1-(4-Hydroxy-2-methylphenyl)ethanone

A188675-100G A188675-25G
77287-446EA 421.27 USD
77287-446 77748-214
4'-Hydroxy-2'-methylacetophenone 97%
4'-Hydroxy-2'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002303
CAS Number: 875-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR