3-Fluoro-4-iodoaniline 98%

Supplier: Ambeed
Warning

A188655-100G A188655-10G A188655-1G A188655-250MG A188655-25G A188655-5G
77287-434EA 341.96 USD
77287-434 77287-436 77287-438 77287-440 77287-442 77287-444
3-Fluoro-4-iodoaniline 98%
3-Fluoro-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.01 g/mol
MDL Number: MFCD07774187
CAS Number: 656-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR