2-Acetyl-3,5(6)-dimethylpyrazine 98% mix

Supplier: Ambeed
Warning

Synonyms: 2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers

A188614-1G A188614-5G
77236-786EA 7.21 USD
77236-786 77236-788
2-Acetyl-3,5(6)-dimethylpyrazine 98% mix
2-Acetyl-3,5(6)-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₄O₂
MW: 300.36 g/mol
MDL Number: MFCD29036871
CAS Number: 72797-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR