10074-G5 98%

Supplier: Ambeed
Danger

Synonyms: N-[1,1′-Biphenyl-2-yl]-7-nitro-2,1,3-Benzoxadiazol-4-amine, c-Myc Inhibitor II, N-([1,1'-Biphenyl]-2-yl)-7-nitrobenzo[c][1,2,5]oxadiazol-4-amine, Biphenyl-2-yl-(7-nitrobenzo[1,2,5]oxadiazol-4-yl)amine, N-2-Biphenylyl-7-nitro-2,1,3-benzoxadiazol-4-amine

A188479-100MG A188479-10MG A188479-1MG A188479-25MG A188479-50MG A188479-5MG
77260-454EA 195 USD
77260-454 77260-456 77260-458 77260-460 77260-462 77260-464
10074-G5 98%
10074-G5

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₄O₃
MW: 332.31 g/mol
MDL Number: MFCD00576774
CAS Number: 413611-93-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR