4-(Trifluoromethyl)salicylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-(trifluoromethyl)benzoic acid

A188438-1000G A188438-100G A188438-10G A188438-25G A188438-500G A188438-5G
77274-348EA 275 USD
77274-348 77274-350 77274-352 77274-354 77274-356 77274-358
4-(Trifluoromethyl)salicylic acid 97%
4-(Trifluoromethyl)salicylic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MDL Number: MFCD01529657
CAS Number: 328-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR