3-Bromo-9,9'-spirobi[fluorene] 98%

Supplier: Ambeed
Warning

A188263-100MG A188263-1G A188263-250MG A188263-5G A188263-10G
77236-766EA 5 USD
77236-766 77236-768 77236-770 77236-772 77746-209
3-Bromo-9,9'-spirobi[fluorene] 98%
3-Bromo-9,9'-spirobi[fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅Br
MDL Number: MFCD28137969
CAS Number: 1361227-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR