3-Iodo-5-nitropyridin-2-ol 97%

Supplier: Ambeed
Warning

A188069-10G A188069-1G A188069-5G
77260-438EA 50.47 USD
77260-438 77260-440 77260-442
3-Iodo-5-nitropyridin-2-ol 97%
3-Iodo-5-nitropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃IN₂O₃
MW: 266 g/mol
MDL Number: MFCD06008303
CAS Number: 25391-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR