3-Aminophthalhydrazide monosodium salt 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium 5-amino-1,4-dioxo-3,4-dihydro-1H-phthalazin-2-ide, Luminol sodium salt
A187906-100MG
A187906-1G
A187906-1MG
A187906-250MG
A187906-25G
A187906-5G
A187906-100G
77236-754EA
8
USD
77236-754
77236-756
77236-758
77236-760
77236-762
77236-764
77746-206
3-Aminophthalhydrazide monosodium salt 98%
3-Aminophthalhydrazide monosodium salt
Precaución: for Research Use Only
Formula:
C₈H₆N₃NaO₂ MW: 199.15 g/mol |
MDL Number:
MFCD00161491 CAS Number: 20666-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...