Rhodium(II) heptafluorobutryate dimer 98%

Supplier: Ambeed
Warning

A187823-100MG A187823-250MG
77248-476EA 118.45 USD
77248-476 77248-478
Rhodium(II) heptafluorobutryate dimer 98%
Rhodium(II) heptafluorobutryate dimer

Precaución: for Research Use Only

Formula: C₁₆F₂₈O₈Rh₂
MW: 1057.93 g/mol
MDL Number: MFCD00799582
CAS Number: 73755-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR