(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N'-((1S,2S)-Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)benzamide), (S,S)-DACH-Phenyl Trost Ligand
A187746-100MG
A187746-1G
A187746-5G
A187746-250MG
A187746-25G
77248-470EA
16.48
USD
77248-470
77248-472
77248-474
77615-588
77746-204
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) 97%
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl)
Precaución: for Research Use Only
Formula:
C₄₄H₄₀N₂O₂P₂ MW: 690.7481 g/mol |
MDL Number:
MFCD01631274 CAS Number: 169689-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...