3-(4-Chlorophenyl)-1,4-dioxane-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A187720-1G A187720-250MG
77236-750EA 275.01 USD
77236-750 77236-752
3-(4-Chlorophenyl)-1,4-dioxane-2-carboxylic acid 95%
3-(4-Chlorophenyl)-1,4-dioxane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClO₄
MW: 242.65 g/mol
CAS Number: 1706443-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR