Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate 97%

Supplier: Ambeed
Danger

A187691-1G A187691-250MG A187691-5G
77248-464EA 21 USD
77248-464 77248-466 77248-468
Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate 97%
Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
MDL Number: MFCD01743485
CAS Number: 35260-93-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR