4-(4-Cyano-2-(2-(4-fluoronaphthalen-1-yl)propanamido)phenyl)butanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: ONO-AE3-208

A187661-10MG A187661-1MG A187661-2MG A187661-50MG A187661-5MG
77236-740EA 63.86 USD
77236-740 77236-742 77236-744 77236-746 77236-748
4-(4-Cyano-2-(2-(4-fluoronaphthalen-1-yl)propanamido)phenyl)butanoic acid 98%
4-(4-Cyano-2-(2-(4-fluoronaphthalen-1-yl)propanamido)phenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₁FN₂O₃
MW: 404.43 g/mol
MDL Number: MFCD13184814
CAS Number: 402473-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR